missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium acetylide, 18 wt.% slurry in xylene, AcroSeal™, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC430701000

 View more versions of this product

Catalog No. AC430701000

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

Sodium acetylide
Low Melting Solid or Crystalline Powder
C2HNa
48.02
1.1990g/mL
White to pale yellow
1066-26-8,1330-20-7
17.0 to 22.0 wt% (Total base)
HCidentical toCNa
MFCD00008568
AcroSeal™ Glass Bottle
100mL

Chemical Identifiers

1066-26-8,1330-20-7
HCidentical toCNa
MFCD00008568
17.0 to 22.0 wt% (Total base)
C2HNa
48.02
Sodium acetylide

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Harmful if inhaled.
Harmful in contact with skin.
Causes skin irritation.
Flammable liquid and vapor.
In contact with water releases flammable gases which may ignite spontaneously.
Reacts violently with water.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Store in a dry place. Store in a closed container.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
Take precautionary measures against static discharge.
Store in a well-ventilated place. Keep cool.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger