Please login to your online account to display your discounted pricing

Sodium acetylide, 18 wt.% slurry in xylene, AcroSeal™, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Sodium acetylide
CAS 1066-26-8
CAS 1330-20-7
Assay 17.0 to 22.0 wt% (Total base)
Molecular Formula C2HNa
Linear Formula HCidentical toCNa
Formula Weight 48.02
MDL Number MFCD00008568
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC430701000

  SDS
thermo fisher scientific
AC430701000
AcroSeal™ Glass Bottle 100mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC430708000

  SDS
thermo fisher scientific
AC430708000
AcroSeal™ Glass Bottle 800mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Low Melting Solid or Crystalline Powder
Formula Weight 48.02
Quantity 100mL
Color White to pale yellow
Packaging AcroSeal™ Glass Bottle
Assay 17.0 to 22.0 wt% (Total base)
Density 1.1990g/mL
Health Hazard 2 GHS H Statement:
Harmful if inhaled.
Harmful in contact with skin.
Causes skin irritation.
Flammable liquid and vapor.
In contact with water releases flammable gases which may ignite spontaneously.
Reacts violently with water.
CAS 1330-20-7
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Store in a dry place. Store in a closed container.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
Take precautionary measures against static discharge.
Store in a well-ventilated place. Keep cool.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material Sodium acetylide
Molecular Formula C2HNa
Linear Formula HCidentical toCNa
MDL Number MFCD00008568
CAS 1066-26-8
Health Hazard 1 GHS Signal Word: Danger
Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Store in a dry place. Store in a closed container.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
Take precautionary measures against static discharge.
Store in a well-ventilated place. Keep cool.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if inhaled.
Harmful in contact with skin.
Causes skin irritation.
Flammable liquid and vapor.
In contact with water releases flammable gases which may ignite spontaneously.
Reacts violently with water.