Please login to your online account to display your discounted pricing

Sodium acetylide, 18 wt.% slurry in xylene, AcroSeal™, ACROS Organics™


Quantity 800mL
Packaging AcroSeal™ Glass Bottle

Manufacturer: thermo fisher scientific  AC430708000

 View more versions of this product

Catalog No. AC430708000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 48.02
Physical Form Low Melting Solid or Crystalline Powder
Quantity 800mL
Color White to pale yellow
Packaging AcroSeal™ Glass Bottle
Assay 17.0 to 22.0 wt% (Total base)
Density 1.1990g/mL
CAS 1330-20-7
Health Hazard 2 GHS H Statement:
Harmful if inhaled.
Harmful in contact with skin.
Causes skin irritation.
Flammable liquid and vapor.
In contact with water releases flammable gases which may ignite spontaneously.
Reacts violently with water.
Health Hazard 1 GHS Signal Word: Danger
Chemical Name or Material Sodium acetylide
CAS 1066-26-8
Molecular Formula C2HNa
MDL Number MFCD00008568
Linear Formula HCidentical toCNa
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Store in a dry place. Store in a closed container.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
Take precautionary measures against static discharge.
Store in a well-ventilated place. Keep cool.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material Sodium acetylide
CAS 1066-26-8
CAS 1330-20-7
Assay 17.0 to 22.0 wt% (Total base)
Molecular Formula C2HNa
Linear Formula HCidentical toCNa
Formula Weight 48.02
MDL Number MFCD00008568
Accessories

GHS Signal Word: Danger

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Store in a dry place. Store in a closed container.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
Take precautionary measures against static discharge.
Store in a well-ventilated place. Keep cool.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if inhaled.
Harmful in contact with skin.
Causes skin irritation.
Flammable liquid and vapor.
In contact with water releases flammable gases which may ignite spontaneously.
Reacts violently with water.

SDS & Safety Documents