Please login to your online account to display your discounted pricing

Sodium acetylide, 18 wt.% slurry in xylene, ACROS Organics™

$59.36 - $59.36

Chemical Identifiers

Chemical Name or Material Sodium acetylide
Name Note 18wt% slurry in xylene
CAS 1066-26-8
CAS 1330-20-7
Assay 17.0 to 22.0 wt%
Molecular Formula C2HNa
Linear Formula HCidentical toCNa
Formula Weight 48.02
MDL Number MFCD00008568
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC342035000

  SDS
thermo fisher scientific
AC342035000
Glass bottle 500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC342031000

  SDS
thermo fisher scientific
342031000
Glass bottle 100g Each for $59.36
Description & Specifications

Specifications

Physical Form Slurry
Formula Weight 48.02
Quantity 500g
Color Yellow to orange to brown
Density 1.1990g/mL
Assay 17.0 to 22.0 wt%
Chemical Name or Material Sodium acetylide
Name Note 18wt% slurry in xylene
Molecular Formula C2HNa
MDL Number MFCD00008568
CAS 1066-26-8
Packaging Glass bottle
Health Hazard 2 GHS H Statement:
Harmful if inhaled.
Harmful in contact with skin.
Causes skin irritation.
Flammable liquid and vapor.
In contact with water releases flammable gases which may ignite spontaneously.
Reacts violently with water.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Store in a dry place. Store in a closed container.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
Take precautionary measures against static discharge.
Store in a well-ventilated place. Keep cool.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Linear Formula HCidentical toCNa
Health Hazard 1 GHS Signal Word: Danger
CAS 1330-20-7

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Store in a dry place. Store in a closed container.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
Take precautionary measures against static discharge.
Store in a well-ventilated place. Keep cool.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if inhaled.
Harmful in contact with skin.
Causes skin irritation.
Flammable liquid and vapor.
In contact with water releases flammable gases which may ignite spontaneously.
Reacts violently with water.