Learn More

Sodium Acetate Trihydrate (White Crystals/USP/FCC/EP/BP), Fisher Chemical™
Manufacturer: Fisher Chemical S607212
Specifications
Sodium Acetate Trihydrate | |
White | |
8 to 9.5 | |
C2H9NaO5 | |
MFCD00071557 | |
0.05% max. (FCC) | |
CC(=O)[O-].O.O.O.[Na+] | |
136.079 | |
CHEBI:32138 | |
Solid | |
BP/EP/FCC/USP | |
0.001% max. (USP) | |
0.05% max. (USP) | |
2mg/kg max. (FCC) | |
Poly Bottle | |
Pass Test (BP) | |
2ppm max. (BP) | |
50ppm max. (BP) | |
1.45g/cm³ |
6131-90-4 | |
58°C | |
2.5kg | |
CH3COONa·3H2O | |
acetic acid, sodium salt, trihydrate, natrium acetate-3-wasser, plasmafusin, sodium acetate tn, sodium acetate trihydrate, sodium acetate usan:jan, sodium acetate usp:jan, thomaegelin, tutofusin, unii-4550k0sc9b | |
AYRVGWHSXIMRAB-UHFFFAOYSA-M | |
sodium;acetate;trihydrate | |
23665404 | |
136.08 | |
99.0 to 101.0% (FCC) | |
0.001% max. (USP), 10mg/kg max. (FCC), 10ppm max. (EP/BP) | |
Pass Test (BP) | |
10ppm max. (BP) | |
36.0 to 41.0% (FCC) | |
Pass Test (FCC) | |
Pass Test (BP) | |
0.005% max. (USP) | |
0.035% max. (USP) |
Chemical Identifiers
6131-90-4 | |
136.079 | |
AYRVGWHSXIMRAB-UHFFFAOYSA-M | |
23665404 | |
sodium;acetate;trihydrate |
C2H9NaO5 | |
MFCD00071557 | |
acetic acid, sodium salt, trihydrate, natrium acetate-3-wasser, plasmafusin, sodium acetate tn, sodium acetate trihydrate, sodium acetate usan:jan, sodium acetate usp:jan, thomaegelin, tutofusin, unii-4550k0sc9b | |
CHEBI:32138 | |
CC(=O)[O-].O.O.O.[Na+] |
Safety and Handling

CAUTION!
Emergency Overview
May cause eye, skin, and respiratory tract irritation. Hygroscopic. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Do not induce vomiting. Obtain medical attention.
NFPA
Health:1
Flammability:1
Instability:1