missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium Acetate Trihydrate (Granular/USP/FCC), Fisher Chemical

Manufacturer:  Fisher Chemical S6083

 View more versions of this product

Catalog No. S608-3




Specifications

Specifications

Sodium Acetate Trihydrate
0.05% max. (FCC)
CH3COONa·3H2O
acetic acid, sodium salt, trihydrate, natrium acetate-3-wasser, plasmafusin, sodium acetate tn, sodium acetate trihydrate, sodium acetate usan:jan, sodium acetate usp:jan, thomaegelin, tutofusin, unii-4550k0sc9b
CC(=O)[O-].O.O.O.[Na+]
136.079
CHEBI:32138
Solid
FCC/USP
0.035% max. (USP)
0.001% max. (USP)
0.05% max. (USP)
36.0 to 41.0% (FCC)
Pass Test (FCC)
0.005% max. (USP)
58°C
3kg
6131-90-4
C2H9NaO5
MFCD00071557
AYRVGWHSXIMRAB-UHFFFAOYSA-M
sodium;acetate;trihydrate
23665404
136.08
99.0 to 101.0% (FCC)
Pass Test (USP)
1.45g/cm³
Pass Test (FCC)
2mg/kg max. (FCC)
Poly Bottle
Meets Requirements (USP)
White
8 to 9.5

Chemical Identifiers

6131-90-4
136.079
AYRVGWHSXIMRAB-UHFFFAOYSA-M
23665404
sodium;acetate;trihydrate
C2H9NaO5
MFCD00071557
acetic acid, sodium salt, trihydrate, natrium acetate-3-wasser, plasmafusin, sodium acetate tn, sodium acetate trihydrate, sodium acetate usan:jan, sodium acetate usp:jan, thomaegelin, tutofusin, unii-4550k0sc9b
CHEBI:32138
CC(=O)[O-].O.O.O.[Na+]
Safety and Handling

Safety and Handling

CAUTION!

Emergency Overview
May cause eye, skin, and respiratory tract irritation. Hygroscopic. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Do not induce vomiting. Obtain medical attention.

NFPA

Health:1
Flammability:1
Instability:1

SDS Documents