missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium Acetate Trihydrate (Granular/USP/FCC/EP/BP/JP), Fisher Chemical™

Manufacturer:  Fisher Chemical S609500

 View more versions of this product

Catalog No. S609500



Specifications

Specifications

Sodium Acetate Trihydrate
Pass Test (JP)
0.2ppm max. (BP)
MFCD00071557
acetic acid, sodium salt, trihydrate, natrium acetate-3-wasser, plasmafusin, sodium acetate tn, sodium acetate trihydrate, sodium acetate usan:jan, sodium acetate usp:jan, thomaegelin, tutofusin, unii-4550k0sc9b
AYRVGWHSXIMRAB-UHFFFAOYSA-M
Report EU/g (EP)
136.079
CHEBI:32138
Solid
BP/EP/FCC/JP/USP
Pass Test (JP)
Clear and Colorless (JP)
Report EU/mg (USP)
0.001% max. (USP)
0.05% max. (USP)
2mg/kg max. (FCC)
Poly Bottle
Pass Test (BP)
0.005% max. (USP)
58°C
500g
6131-90-4
0.05% max. (FCC)
C2H9NaO5
Pass Test (BP)
2ppm max. (BP)
CC(=O)[O-].O.O.O.[Na+]
sodium;acetate;trihydrate
23665404
136.08
99.0 to 101.0% (EP)
50ppm max. (BP)
0.011% max. (JP)
1.45g/cm³
10ppm max. (BP)
Pass Test (BP)
10ppm max. (BP)
36.0 to 41.0% (FCC)
Pass Test (FCC)
Meets Requirements (BP)
White
7.5 to 9.2 (USP), 7.5 to 9.0 (EP/BP)

Chemical Identifiers

6131-90-4
136.079
AYRVGWHSXIMRAB-UHFFFAOYSA-M
23665404
sodium;acetate;trihydrate
C2H9NaO5
MFCD00071557
acetic acid, sodium salt, trihydrate, natrium acetate-3-wasser, plasmafusin, sodium acetate tn, sodium acetate trihydrate, sodium acetate usan:jan, sodium acetate usp:jan, thomaegelin, tutofusin, unii-4550k0sc9b
CHEBI:32138
CC(=O)[O-].O.O.O.[Na+]
Safety and Handling

Safety and Handling

CAUTION!

Emergency Overview
May cause eye, skin, and respiratory tract irritation. Hygroscopic. Use personal protective equipment. Ensure adequate ventilation. Wash off immediately with plenty of water for at least 15 minutes. Get medical attention immediately if symptoms occur. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Obtain medical attention. Move to fresh air. If breathing is difficult, give oxygen. Get medical attention immediately if symptoms occur. Do not induce vomiting. Do not induce vomiting. Obtain medical attention.

NFPA

Health:1
Flammability:1
Instability:1

SDS Documents