Please login to your online account to display your discounted pricing

Sodium Acetate Trihydrate (Colorless-to-white Crystals), Fisher BioReagents

$59.27 - $84.59

Chemical Identifiers

Chemical Name or Material Sodium Acetate Trihydrate
CAS 6131-90-4
Physical Form Solid
Assay 99.0 to 101.0%
Molecular Formula C2H3NaO2 · 3 H2O
Linear Formula CH3COONa·3H2O
Formula Weight 136.08
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

BP334-1

  SDS
fisher scientific company
BP3341
Poly Bottle 1kg Each for $84.59
Case of 6 Each for $507.54

BP334-500

  SDS
fisher scientific company
BP334500
Poly Bottle 500g Each for $59.27
Case of 6 Each for $355.62
Description & Specifications

Specifications

Melting Point 58°C
Formula Weight 136.08
Quantity 1kg
Assay 99.0 to 101.0%
Physical Form Solid
Molecular Formula C2H3NaO2 · 3 H2O
pH 8 to 9.5
Color White
Density 1.45g/cm³
Phosphate 5ppm max.
Chloride 0.001% max.
Insoluble Matter 0.005% max.
Sulfate 0.002% max.
Heavy Metals (as Pb) 5ppm max.
CAS 6131-90-4
Recommended Storage RT
ChemAlert Storage Symbol Gray
Substances Reducing Permanganate Pass Test
Packaging Poly Bottle
Magnesium (Mg) 0.002% max.
Calcium (Ca) 0.005% max.
Iron (Fe) 5ppm max.
Potassium (K) 0.005% max.
Chemical Name or Material Sodium Acetate Trihydrate
Linear Formula CH3COONa·3H2O

The trihydrate form of Sodium Acetate can be used for many of the same applications as the anhydrous form.

Safety and Handling
 

RT