Learn More
Sodium acetate trihydrate, ACS, 99.0%-101%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 011553A3
Description
In photography, analytical laboratory as buffer and in pharmaceutical.Sodium acetate trihydrate is used in various applications. It is used as a buffer of a pH range in between to 4.0 to 6.0. It is also used in the purification and precipitation of nucleic acids and in protein crystallization. It is used as a mordant dyeing, as a pickling agent in chrome tanning, as a neutralizer in waste streams containing sulfuric acid, in the textile industry and as a photoresist while using aniline dyes. It plays an important role to retard vulcanization of chloroprene in synthetic rubber production. It is a precursor to prepare ester from alkyl halide.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Sodium acetate trihydrate | |
For ACS analysis | |
Crystalline | |
99.0% to 100.5% | |
MFCD00071557 | |
sodium acetate trihydrate, thomaegelin, acetic acid, sodium salt, trihydrate, plasmafusin, tutofusin, natrium acetate-3-wasser, unii-4550k0sc9b, sodium acetate usp:jan, sodium acetate usan:jan, sodium acetate tn | |
AYRVGWHSXIMRAB-UHFFFAOYSA-M | |
sodium;acetate;trihydrate | |
23665404 | |
136.08 (82.03 Anhydrous) | |
1.45 g/mL |
62°C to 64°C | |
6131-90-4 | |
2 kg | |
C2H9NaO5 | |
3732037 | |
Freely soluble in alcohol. Soluble in water and alcohol. | |
CC(=O)[O-].O.O.O.[Na+] | |
136.079 | |
CHEBI:32138 | |
ACS Reagent |
Chemical Identifiers
6131-90-4 | |
136.079 | |
AYRVGWHSXIMRAB-UHFFFAOYSA-M | |
23665404 | |
sodium;acetate;trihydrate |
C2H9NaO5 | |
MFCD00071557 | |
sodium acetate trihydrate, thomaegelin, acetic acid, sodium salt, trihydrate, plasmafusin, tutofusin, natrium acetate-3-wasser, unii-4550k0sc9b, sodium acetate usp:jan, sodium acetate usan:jan, sodium acetate tn | |
CHEBI:32138 | |
CC(=O)[O-].O.O.O.[Na+] |
Safety and Handling
H500
EINECSNumber : 204-823-8
RTECSNumber : AJ4580000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only