Please login to your online account to display your discounted pricing

Sodium Acetate, Trihydrate, ACS Grade, 99.0% to 101.0%, LabChem™

$71.91 - $71.91

Chemical Identifiers

Chemical Name or Material Sodium Acetate
CAS 6131-90-4
CAS Max % 100
Name Note Trihydrate
Purity Grade ACS Grade
Linear Formula CH3COOHNa·3H2O
Formula Weight 136.08
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC228201

 
lab chem
LC228201
500g Each for $71.91
Description & Specifications

Specifications

Assay 99.0 to 101.0%
Formula Weight 136.08
Chemical Name or Material Sodium Acetate
Name Note Trihydrate
Reducing Substances Passes Test
Solubility Soluble in water
Specific Gravity 1.45g/cm3
Boiling Point 123°C
Quantity 500g
Health Hazard 3 GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Identification Passes Test
Packaging Poly Bottle
Purity Grade ACS Grade
Physical Form Crystals
Storage Information Room Temperature
Linear Formula CH3COOHNa·3H2O
CAS 6131-90-4
Decomposition Information Fume; Carbon monoxide; Carbon dioxide
Color White
Melting Point 58°C
CAS Max % 100
Health Hazard 2 GHS H Statement
Product is not hazardous.
Density 1.45g/cm3
Safety and Handling

GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.

GHS H Statement
Product is not hazardous.