Please login to your online account to display your discounted pricing

Sodium Acetate Trihydrate ACS, EMD Millipore

Indifferent to potassium permanganate

$313.26 - $313.26

Chemical Identifiers

Chemical Name or Material Sodium Acetate
CAS 6131-90-4
Name Note EMSURE™, For Analysis
Purity Grade ACS Grade, ISO, Reagent Grade, EP
Molecular Formula C2H3NaO2 · 3H2O
Linear Formula CH3COONa · 3H2O
Formula Weight 136.8
View More Specs
Products
Catalog Number Mfr. No. Price Quantity    

MSX02553

 
emd millipore corporation
SX02553
Each for $313.26
Description & Specifications

Specifications

Chemical Name or Material Sodium Acetate
Name Note EMSURE™, For Analysis
CAS 6131-90-4
Recommended Storage Store without restrictions
Purity Grade ACS Grade, ISO, Reagent Grade, EP
Molecular Formula C2H3NaO2 · 3H2O
Linear Formula CH3COONa · 3H2O
Formula Weight 136.8