Please login to your online account to display your discounted pricing

Sodium Acetate, For Arsenic, Certified, 0.200M ±0.002M (2.0N), LabChem™

$77.06 - $77.06

Chemical Identifiers

Chemical Name or Material Sodium Acetate
CAS 7732-18-5
CAS Max % 97.3
CAS 6131-90-4
CAS Max % 2.7
Name Note For Arsenic
Purity Grade Certified Grade
Linear Formula CH3COOHNa·3H2O
Formula Weight 136.08
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

LC228401

 
lab chem
LC228401
500mL Each for $77.06
Description & Specifications

Specifications

Purity Grade Certified Grade
Formula Weight 136.08
Chemical Name or Material Sodium Acetate
Traceability to NIST Traceable to NIST
Viscosity 1.1cSt
Decomposition Information Fume; Carbon monoxide; Carbon dioxide
Color Clear, colorless
Quantity 500mL
Name Note For Arsenic
Physical Form Liquid
Concentration 0.200M ±0.002M (2.0N)
Linear Formula CH3COOHNa·3H2O
Density 1.01g/mL
CAS 7732-18-5
CAS Max % 97.3
Health Hazard 2 GHS H Statement
Solution is not hazardous.
Identification Passes Test
CAS 6131-90-4
Health Hazard 3 GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Packaging Poly Bottle
CAS Max % 2.7
Solubility Soluble in water
Specific Gravity 1.01g/mL
Storage Information Room Temperature
Safety and Handling

GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.

GHS H Statement
Solution is not hazardous.