missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Sodium Acetate, Anhydrous, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™
High quality chemicals for laboratory and specialized industrial use
Supplier: Avantor J.T.Baker 347001
Specifications
Sodium Acetate | |
127-09-3 | |
C2H3NaO2 | |
sodium acetate, acetic acid, sodium salt, sodium acetate anhydrous, sodium acetate, anhydrous, acetic acid sodium salt, anhydrous sodium acetate, sodii acetas, sodium ethanoate, natrium aceticum, octan sodny czech | |
[Na+].CC([O-])=O | |
82.03 | |
CHEBI:32954 | |
BAKER ANALYZED™ ACS Reagent | |
1L = 1.53kg |
Anhydrous | |
500 g | |
MFCD00012459 | |
VMHLLURERBWHNL-UHFFFAOYSA-M | |
sodium;acetate | |
517045 | |
82.03 | |
Poly Bottle |
Chemical Identifiers
127-09-3 | |
82.03 | |
VMHLLURERBWHNL-UHFFFAOYSA-M | |
517045 | |
[Na+].CC([O-])=O |
C2H3NaO2 | |
MFCD00012459 | |
sodium acetate, acetic acid, sodium salt, sodium acetate anhydrous, sodium acetate, anhydrous, acetic acid sodium salt, anhydrous sodium acetate, sodii acetas, sodium ethanoate, natrium aceticum, octan sodny czech | |
CHEBI:32954 |