Please login to your online account to display your discounted pricing

Sodium acetate, anhydrous, 99.997% (metals basis), Alfa Aesar™

Chemical Identifiers

Chemical Name or Material Sodium acetate
CAS 127-09-3
Name Note anhydrous
Molecular Formula NaOOCCH3
Formula Weight 82.03
MDL Number MFCD00012459
Beilstein 3595639
Merck Index 14,8571
Synonym Acetic acid sodium salt
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AA1100522

 
alfa aesar
1100522
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 82.03
Quantity 100g
Chemical Name or Material Sodium acetate
TSCA TSCA
Name Note anhydrous
Physical Form Powder
Assay 99.997%
Sensitivity Hygroscopic
Assay Percent Notes (metals basis)
Molecular Formula NaOOCCH3
MDL Number MFCD00012459
CAS 127-09-3
Beilstein 3595639
Merck Index 14,8571
Synonym Acetic acid sodium salt
Melting Point 324°C
pH 7.5 to 9
Density 1.528
RTECS Number AJ4300010