Learn More
Sodium acetate, 99+%, anhydrous, Molecular Biology Reagent, Thermo Scientific Chemicals
$301.73 - $301.73
Chemical Identifiers
CAS | 127-09-3 |
---|---|
Molecular Formula | C2H3NaO2 |
Molecular Weight (g/mol) | 82.03 |
MDL Number | MFCD00012459 |
InChI Key | VMHLLURERBWHNL-UHFFFAOYSA-M |
Synonym | sodium acetate, acetic acid, sodium salt, sodium acetate anhydrous, sodium acetate, anhydrous, acetic acid sodium salt, anhydrous sodium acetate, sodii acetas, sodium ethanoate, natrium aceticum, octan sodny czech |
PubChem CID | 517045 |
ChEBI | CHEBI:32954 |
SMILES | [Na+].CC([O-])=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAJ6639736
|
Thermo Scientific Chemicals
J6639736 |
500 g |
Each for $301.73
|
|
|||||
Description
Sodium acetate, 99+%, anhydrous, is been widely used in for precipitation of nucleic acids and preparation of gel stains for protein gel electrophoresis. Sodium acetate buffer solution is suitable for precipitation of DNA, elution of proteins and for use in molecular biology applications such as ChIP-on-ChIP assay and in vitro enzyme inhibition assay.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsSodium acetate, 99+%, anhydrous, is been widely used in for precipitation of nucleic acids and preparation of gel stains for protein gel electrophoresis. Sodium acetate buffer solution is suitable for precipitation of DNA, elution of proteins and for use in molecular biology applications such as ChIP-on-ChIP assay and in vitro enzyme inhibition assay.
Solubility
Soluble in water (1.19 g/L) at 20°C, and ethanol.
Notes
Hygroscopic. Protect from heat. Store under dry inert gas. Incompatible with moisture and oxidizing agents.
Chemical Identifiers
127-09-3 | |
82.03 | |
VMHLLURERBWHNL-UHFFFAOYSA-M | |
517045 | |
[Na+].CC([O-])=O |
C2H3NaO2 | |
MFCD00012459 | |
sodium acetate, acetic acid, sodium salt, sodium acetate anhydrous, sodium acetate, anhydrous, acetic acid sodium salt, anhydrous sodium acetate, sodii acetas, sodium ethanoate, natrium aceticum, octan sodny czech | |
CHEBI:32954 |
Specifications
324°C | |
White | |
Powder | |
C2H3NaO2 | |
>110°C (230°F) | |
sodium acetate, acetic acid, sodium salt, sodium acetate anhydrous, sodium acetate, anhydrous, acetic acid sodium salt, anhydrous sodium acetate, sodii acetas, sodium ethanoate, natrium aceticum, octan sodny czech | |
VMHLLURERBWHNL-UHFFFAOYSA-M | |
sodium;acetate | |
517045 | |
82.03 | |
Hygroscopic |
127-09-3 | |
7.5 to 9 | |
500 g | |
MFCD00012459 | |
14,8571 | |
Soluble in water (1.19g/L) at 20°C,and ethanol. | |
[Na+].CC([O-])=O | |
82.03 | |
CHEBI:32954 | |
≥99% | |
Sodium acetate, Anhydrous, Molecular Biology Reagent |
Safety and Handling
H500
EINECSNumber : 204-823-8
RTECSNumber : AJ4300010
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Argon
RUO – Research Use Only