Please login to your online account to display your discounted pricing

Sodium Acetate, 50% (w/v), Ricca Chemical

Chemical Identifiers

Chemical Name or Material Sodium Acetate
CAS 6131-90-4
CAS 7732-18-5
View More Specs
Products
Catalog Number Mfr. No. Price Quantity    

R71207001A

 
ricca chemical company
RIR71207001A
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

R71207004A

 
ricca chemical company
RIR71207004A
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Liquid
Color Clear colorless
Shelf Life 24 months
Packaging Poly Bottle
Assay Percent Range 50% (w/v)
CAS 6131-90-4
Odor odorless to slight acetic acid odor
CAS 7732-18-5
Chemical Name or Material Sodium Acetate
Safety and Handling

24 months