Please login to your online account to display your discounted pricing

Alfa Aesar™ Sodium acetate, 3M aq. soln., pH 5.2, autoclaved

Chemical Identifiers

Chemical Name or Material Sodium acetate
CAS 127-09-3
Name Note Autoclaved
Linear Formula NaOOCCH3
Formula Weight 82.03
MDL Number MFCD00012459
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ63560EQE

  SDS
alfa aesar™
J63560EQE
125mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 82.03
Chemical Name or Material Sodium acetate
CAS 127-09-3
pH 5.2
Name Note Autoclaved
Linear Formula NaOOCCH3
Odor Odorless
TSCA TSCA
Quantity 125mL
Physical Form Liquid
Concentration 3M aq. soln.
MDL Number MFCD00012459