Please login to your online account to display your discounted pricing

Sodium Acetate, 20% (w/w) Aqueous Solution, Ricca Chemical

Chemical Identifiers

Chemical Name or Material Sodium Acetate
CAS 7732-18-5
CAS Min % 78.0
CAS Max % 82.0
CAS 6131-90-4
CAS Min % 19.61
CAS Max % 20.41
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

71204

 
ricca chemical company
RI71204
Poly natural 120mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

R712000020F

 
ricca chemical company
RIR712000020F
Cubitainer 20L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Quantity 120mL
Chemical Name or Material Sodium Acetate
CAS 7732-18-5
CAS Max % 82.0
Concentration or Composition Notes 20% (w/w)
Solvent or Matrix Aqueous Solution
CAS 6131-90-4
CAS Min % 19.61
CAS Max % 20.41
CAS Min % 78.0
Packaging Poly natural