missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Sodium Acetate, 20% (w/w), Ricca Chemical
APHA for Iron Testing
Supplier: Ricca Chemical Company R712000020F
Specifications
Sodium Acetate | |
78.0,19.61 | |
C2H9NaO5 | |
AYRVGWHSXIMRAB-UHFFFAOYSA-M | |
sodium;acetate;trihydrate | |
23665404 | |
Cubitainer | |
20 L |
6131-90-4,7732-18-5 | |
82.0,20.41 | |
sodium acetate trihydrate, thomaegelin, acetic acid, sodium salt, trihydrate, plasmafusin, tutofusin, natrium acetate-3-wasser, unii-4550k0sc9b, sodium acetate usp:jan, sodium acetate usan:jan, sodium acetate tn | |
CC(=O)[O-].O.O.O.[Na+] | |
136.079 | |
CHEBI:32138 | |
Aqueous Solution | |
20% (w/w) |
Chemical Identifiers
Sodium Acetate |