missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Sodium acetate, 1M aq. soln., pH 4.5, RNAse free

Manufacturer:  Alfa Aesar™ J63669AK

 View more versions of this product

Catalog No. AAJ63669AK



Description

Description

Applications
Sodium Acetate is used as a buffer in the buffering range of pH 3.6-5.6. And also used in the purification and precipitation of nucleic acids, in protein crystallization, Staining of gels in protein gel electrophoresis and HPLC. Mordant dyeing; DNA microarray studies of E. coli response. Investigation of protein unfolding during reverse phase chromatography. Used in conjunction with sodium carbonate to enhance the activation of freeze-dried subtilisin Carlsberg in organic solvents. As a substrate of acetokinase.

Solubility
Fully miscible.

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications

Specifications

Sodium acetate
127-09-3
NaOOCCH3
acetic acid sodium salt, acetic acid, sodium salt, anhydrous sodium acetate, natrium aceticum, octan sodny czech, sodii acetas, sodium acetate, sodium acetate anhydrous, sodium acetate, anhydrous, sodium ethanoate
VMHLLURERBWHNL-UHFFFAOYSA-M
sodium;acetate
517045
82.03
1M aq. soln.
4.5
RNAse free
C2H3NaO2
MFCD00012459
Fully miscible.
CC(=O)[O-].[Na+]
82.034
CHEBI:32954
Liquid
Odorless
250mL

Chemical Identifiers

127-09-3
82.034
VMHLLURERBWHNL-UHFFFAOYSA-M
517045
sodium;acetate
C2H3NaO2
MFCD00012459
acetic acid sodium salt, acetic acid, sodium salt, anhydrous sodium acetate, natrium aceticum, octan sodny czech, sodii acetas, sodium acetate, sodium acetate anhydrous, sodium acetate, anhydrous, sodium ethanoate
CHEBI:32954
CC(=O)[O-].[Na+]
Safety and Handling

Safety and Handling

TSCA : Yes

Recommended Storage : Ambient temperatures

Documents