Please login to your online account to display your discounted pricing

Sodium Acetate, 0.2M, APHA for Arsenic, Ricca Chemical

$58.36 - $58.36

Chemical Identifiers

Chemical Name or Material Sodium Acetate
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

7129-16

  SDS
ricca chemical company
RI712916
Plastic 500mL Each for $58.36

7129-1

  SDS
ricca chemical company
RI71291
Plastic 4L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

7129-32

  SDS
ricca chemical company
RI712932
Plastic 1L CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Sodium Acetate
Packaging Plastic
Methods APHA for Arsenic
Quantity 500mL
Concentration or Composition (by Analyte or Components) 0.200M (M/5)