Please login to your online account to display your discounted pricing

Sodium AA Standard, 1mL = 1mg Na (1,000ppm Na)NaCl in Water, Ricca Chemical

$59.73 - $160.64

Chemical Identifiers

Chemical Name or Material Sodium AA Standard
CAS 7732-18-5
CAS Min % 98.0
CAS Max % 100.0
CAS 7647-14-5
CAS Min % 0.25
CAS Max % 0.26
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

ANA1KW100

 
ricca chemical company
RIANA1KW100
Poly natural 100mL Each for $59.73

ANA1KW500

 
ricca chemical company
RIANA1KW500
Poly natural 500mL Each for $160.64
Description & Specifications

Specifications

Quantity 100mL
Color Clear colorless
Odor Odorless
Chemical Name or Material Sodium AA Standard
Shelf Life 24 months
CAS 7732-18-5
Concentration or Composition (by Analyte or Components) 1mL = 1mg Na (1, 000ppm Na)NaCl in Water
CAS Min % 98.0
CAS Max % 100.0
CAS 7647-14-5
Packaging Poly natural
CAS Max % 0.26
CAS Min % 0.25
Safety and Handling

24 months