missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Sodium 8-Ethoxyquinoline-5-sulfonate Hydrate 98.0+%, TCI America™
$114.48 - $336.78
Chemical Identifiers
CAS | 7246-07-3 |
---|---|
Molecular Formula | C11H10NNaO4S |
Molecular Weight (g/mol) | 275.25 |
MDL Number | MFCD00006794 |
InChI Key | KGBPIIYKHSGTAJ-UHFFFAOYSA-M |
Synonym | Actinoquinol Sodium Salt, 8-Ethoxyquinoline-5-sulfonic Acid Sodium Salt |
PubChem CID | 87469405 |
IUPAC Name | 8-ethoxyquinoline-5-sulfonic acid;sodium |
SMILES | CCOC1=C2C(=C(C=C1)S(=O)(=O)O)C=CC=N2.[Na] |
Chemical Identifiers
7246-07-3 | |
275.25 | |
KGBPIIYKHSGTAJ-UHFFFAOYSA-M | |
87469405 | |
CCOC1=C2C(=C(C=C1)S(=O)(=O)O)C=CC=N2.[Na] |
C11H10NNaO4S | |
MFCD00006794 | |
Actinoquinol Sodium Salt, 8-Ethoxyquinoline-5-sulfonic Acid Sodium Salt | |
8-ethoxyquinoline-5-sulfonic acid;sodium |
Specifications
7246-07-3 | |
C11H10NNaO4S | |
5 g | |
KGBPIIYKHSGTAJ-UHFFFAOYSA-M | |
8-ethoxyquinoline-5-sulfonic acid;sodium | |
87469405 | |
≥98.0% (HPLC) | |
Sodium 8-Ethoxyquinoline-5-sulfonate Hydrate |
White | |
MFCD00006794 | |
Actinoquinol Sodium Salt, 8-Ethoxyquinoline-5-sulfonic Acid Sodium Salt | |
CCOC1=C2C(=C(C=C1)S(=O)(=O)O)C=CC=N2.[Na] | |
275.25 | |
275.25 | |
Crystalline Powder |
Safety and Handling
TSCA : No