Learn More
Sodium 4-hydroxybenzenesulfonate dihydrate, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1480836
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Sodium 4-hydroxybenzenesulfonate dihydrate | |
>300°C | |
C6H5O4S | |
500 g | |
sodium 4-hydroxybenzenesulfonate dihydrate, unii-5nh81n759q, phenol-4-sulfonic acid sodium salt dihydrate, 4-hydroxybenzenesulfonic acid sodium salt dihydrate, benzenesulfonic acid, 4-hydroxy-, monosodium salt, dihydrate, 4-hydroxybenzenesulfonic acid sodium salt, sodium 4-hydroxybenzene-1-sulfonate dihydrate, phenolsulphonate sodium usp, acmc-2098hs, sodium p-phenolsulfonate dihydrate | |
OC1=CC=C(C=C1)S([O-])(=O)=O | |
173.16 | |
232.20 (196.17 Anhydrous) |
10580-19-5 | |
Odorless | |
MFCD00044734,MFCD00150724 | |
4040891 | |
FEPBITJSIHRMRT-UHFFFAOYSA-M | |
sodium;4-hydroxybenzenesulfonate;dihydrate | |
23666329 | |
97% |
Chemical Identifiers
10580-19-5 | |
173.16 | |
FEPBITJSIHRMRT-UHFFFAOYSA-M | |
23666329 | |
OC1=CC=C(C=C1)S([O-])(=O)=O |
C6H5O4S | |
MFCD00044734,MFCD00150724 | |
sodium 4-hydroxybenzenesulfonate dihydrate, unii-5nh81n759q, phenol-4-sulfonic acid sodium salt dihydrate, 4-hydroxybenzenesulfonic acid sodium salt dihydrate, benzenesulfonic acid, 4-hydroxy-, monosodium salt, dihydrate, 4-hydroxybenzenesulfonic acid sodium salt, sodium 4-hydroxybenzene-1-sulfonate dihydrate, phenolsulphonate sodium usp, acmc-2098hs, sodium p-phenolsulfonate dihydrate | |
sodium;4-hydroxybenzenesulfonate;dihydrate |
Safety and Handling
GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 212-550-0
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only