missing translation for 'onlineSavingsMsg'
Learn More

Sodium 4-(2-Bromoethyl)benzenesulfonate 98.0+%, TCI America™

Supplier:  TCI America S048425G

 View more versions of this product

Catalog No. S048425G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Sodium 4-(2-Bromoethyl)benzenesulfonate
White
MFCD06797167
4-(2-Bromoethyl)benzenesulfonic Acid Sodium Salt
C1=CC(=CC=C1CCBr)S(=O)(=O)[O-].[Na+]
287.103
287.10
Crystalline Powder
65036-65-9
C8H8BrNaO3S
25 g
ZFGDZMNLHTUEAV-UHFFFAOYSA-M
sodium;4-(2-bromoethyl)benzenesulfonate
23677320
≥98.0% (T)

Chemical Identifiers

65036-65-9
287.103
ZFGDZMNLHTUEAV-UHFFFAOYSA-M
23677320
C1=CC(=CC=C1CCBr)S(=O)(=O)[O-].[Na+]
C8H8BrNaO3S
MFCD06797167
4-(2-Bromoethyl)benzenesulfonic Acid Sodium Salt
sodium;4-(2-bromoethyl)benzenesulfonate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions