missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium 3-Nitrobenzoate, TCI America™

Manufacturer:  TCI America S0837500G

 View more versions of this product

Catalog No. S0837500G



Specifications

Specifications

Sodium 3-Nitrobenzoate
500g
C7H4NNaO4
sodium 3-nitrobenzoate, 3-nitrobenzoic acid sodium salt, benzoic acid, 3-nitro-, sodium salt, unii-i1eq837b3j, m-nitrobenzoic acid sodium salt, sodium m-nitrobenzoate, benzoic acid, 3-nitro-, sodium salt 1:1, 3-nitrobenzoic acid, sodium salt, acmc-209pqe, dsstox_cid_24733
C1=CC(=CC(=C1)[N+](=O)[O-])C(=O)[O-].[Na+]
189.102
189.10
≥98.0% (HPLC,T)
White-Yellow
827-95-2
MFCD00051097
MUADFEZFSKAZLT-UHFFFAOYSA-M
sodium;3-nitrobenzoate
70014
Crystal-Powder at 20°C

Chemical Identifiers

827-95-2
189.102
MUADFEZFSKAZLT-UHFFFAOYSA-M
70014
C1=CC(=CC(=C1)[N+](=O)[O-])C(=O)[O-].[Na+]
C7H4NNaO4
MFCD00051097
sodium 3-nitrobenzoate, 3-nitrobenzoic acid sodium salt, benzoic acid, 3-nitro-, sodium salt, unii-i1eq837b3j, m-nitrobenzoic acid sodium salt, sodium m-nitrobenzoate, benzoic acid, 3-nitro-, sodium salt 1:1, 3-nitrobenzoic acid, sodium salt, acmc-209pqe, dsstox_cid_24733
sodium;3-nitrobenzoate
Safety and Handling

Safety and Handling

EINECSNumber : (3)-3027

TSCA : Yes

Documents