Learn More
Sodium 3-nitrobenzenesulfonate, 98%, water <2%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A102440E
Description
Sodium 3-nitrobenzenesulfonate is used in the synthesis of quinoline. It is also used in Stabilizer for dyeing of fibers; assistant in discharge printing; oxidizing agent in demetalizers and industrial cleaners. It is also used as an developing agent for electroplating and auxiliary for dying fabrics.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsSodium 3-nitrobenzenesulfonate is used in the synthesis of quinoline. It is also used in Stabilizer for dyeing of fibers; assistant in discharge printing; oxidizing agent in demetalizers and industrial cleaners. It is also used as an developing agent for electroplating and auxiliary for dying fabrics.
Solubility
Soluble in water 200 g/L (20C).
Notes
Hygroscopic store in inert gas Store away from strong oxidizing agents. keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications
Sodium 3-nitrobenzenesulfonate | |
127-68-4 | |
C6H4NNaO5S | |
2500 g | |
sodium 3-nitrobenzenesulfonate, 3-nitrobenzenesulfonic acid sodium salt, sodium 3-nitrobenzenesulphonate, sodium m-nitrobenzenesulfonate, nitrol s, tiskan czech, ludigol, ludigol f,60, benzenesulfonic acid, 3-nitro-, sodium salt, unii-1f11sxj4c6 | |
LJRGBERXYNQPJI-UHFFFAOYSA-M | |
sodium;3-nitrobenzenesulfonate | |
31389 | |
98% |
Water <2% | |
350°C | |
MFCD00007490 | |
Hygroscopic | |
Soluble in water 200g/L (20C). | |
C1=CC(=CC(=C1)S(=O)(=O)[O-])[N+](=O)[O-].[Na+] | |
225.15 | |
225.16 (Anhydrous) |
Chemical Identifiers
127-68-4 | |
225.15 | |
LJRGBERXYNQPJI-UHFFFAOYSA-M | |
31389 | |
C1=CC(=CC(=C1)S(=O)(=O)[O-])[N+](=O)[O-].[Na+] |
C6H4NNaO5S | |
MFCD00007490 | |
sodium 3-nitrobenzenesulfonate, 3-nitrobenzenesulfonic acid sodium salt, sodium 3-nitrobenzenesulphonate, sodium m-nitrobenzenesulfonate, nitrol s, tiskan czech, ludigol, ludigol f,60, benzenesulfonic acid, 3-nitro-, sodium salt, unii-1f11sxj4c6 | |
sodium;3-nitrobenzenesulfonate |
Safety and Handling
GHS H Statement
H319-H317
Causes serious eye irritation.
May cause an allergic skin reaction.
P261-P264b-P272-P280-P302+P352-P305+P351+P338-P333+P313-P363-P501c
H317-H319
EINECSNumber : 204-857-3
RTECSNumber : DB7195000
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only