missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Sodium 3-nitrobenzenesulfonate, 98%, water <2%

Manufacturer:  Alfa Aesar™ A1024436

 View more versions of this product

Catalog No. AAA1024436



Description

Description

Applications
Sodium 3-nitrobenzenesulfonate is used in the synthesis of quinoline. It is also used in Stabilizer for dyeing of fibers; assistant in discharge printing; oxidizing agent in demetalizers and industrial cleaners. It is also used as an developing agent for electroplating and auxiliary for dying fabrics.

Solubility
Soluble in water 200 g/L (20C).

Notes
Hygroscopic store in inert gas Store away from strong oxidizing agents. keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications

Specifications

Sodium 3-nitrobenzenesulfonate
127-68-4
C6H4NNaO5S
3-Nitrobenzenesulfonic acid sodium salt hydrate
C1=CC(=CC(=C1)S(=O)(=O)[O-])[N+](=O)[O-].[Na+]
225.15
225.16 (Anhydrous)
500g
Water <2%
98%
MFCD00007490
LJRGBERXYNQPJI-UHFFFAOYSA-M
sodium;3-nitrobenzenesulfonate
31389
350°C

Chemical Identifiers

127-68-4
225.15
LJRGBERXYNQPJI-UHFFFAOYSA-M
31389
C1=CC(=CC(=C1)S(=O)(=O)[O-])[N+](=O)[O-].[Na+]
C6H4NNaO5S
MFCD00007490
3-Nitrobenzenesulfonic acid sodium salt hydrate
sodium;3-nitrobenzenesulfonate
Safety and Handling

Safety and Handling

GHS H Statement
H319-H317
Causes serious eye irritation.
May cause an allergic skin reaction.

GHS P Statement
P261-P280-P305+P351+P338-P363-P321-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Wash contaminated clothing before reuse.
Specific treatment (see ... on this label).
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

RTECSNumber : DB7195000

TSCA : TSCA

SDS Documents