Please login to your online account to display your discounted pricing

sodium 3,5-dichloro-6-hydroxybenzenesulfonate, Maybridge

Chemical Identifiers

Chemical Name or Material Sodium 3, 5-chloro-6-hydroxybenzenesulfonate
Assay 97%
Assay Percent Range 97%
Molecular Formula C6H3Cl2O4S·Na
Linear Formula Cl2C6H2(OH)SO3Na
Formula Weight 265.05
Synonym HDCBS
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

BTB09289EA

  SDS
fisher scientific company
BTB09289EA
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 265.05
Quantity 10g
Assay 97%
Assay Percent Range 97%
Linear Formula Cl2C6H2(OH)SO3Na
Molecular Formula C6H3Cl2O4S·Na
CAS 54970-72-8
Chemical Name or Material Sodium 3, 5-chloro-6-hydroxybenzenesulfonate
Synonym HDCBS