Please login to your online account to display your discounted pricing

sodium 3,5-dichloro-6-hydroxybenzenesulfonate, Maybridge


Quantity 10g

Manufacturer: fisher scientific company  BTB09289EA

 View more versions of this product

Catalog No. BTB09289EA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 265.05
Quantity 10g
Assay 97%
Chemical Name or Material Sodium 3, 5-chloro-6-hydroxybenzenesulfonate
Assay Percent Range 97%
Molecular Formula C6H3Cl2O4S·Na
Linear Formula Cl2C6H2(OH)SO3Na
CAS 54970-72-8
Synonym HDCBS

Specifications

Chemical Name or Material Sodium 3, 5-chloro-6-hydroxybenzenesulfonate
Assay 97%
Assay Percent Range 97%
Molecular Formula C6H3Cl2O4S·Na
Linear Formula Cl2C6H2(OH)SO3Na
Formula Weight 265.05
Synonym HDCBS