missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium 2-Propylvalerate, TCI America™

Manufacturer:  TCI America S0894100G

 View more versions of this product

Catalog No. S0894100G



Specifications

Specifications

Sodium 2-Propylvalerate
100g
C8H15NaO2
sodium valproate, sodium 2-propylpentanoate, valproate sodium, valproic acid sodium salt, eurekene, labazene, orfiril, epilim, valproic acid sodium, valerin
CCCC(CCC)C(=O)[O-].[Na+]
166.196
CHEBI:9925
Crystal-Powder at 20°C
White
1069-66-5
MFCD00078604
AEQFSUDEHCCHBT-UHFFFAOYSA-M
sodium;2-propylpentanoate
16760703
166.20
≥98.0% (T)

Chemical Identifiers

1069-66-5
166.196
AEQFSUDEHCCHBT-UHFFFAOYSA-M
16760703
sodium;2-propylpentanoate
C8H15NaO2
MFCD00078604
sodium valproate, sodium 2-propylpentanoate, valproate sodium, valproic acid sodium salt, eurekene, labazene, orfiril, epilim, valproic acid sodium, valerin
CHEBI:9925
CCCC(CCC)C(=O)[O-].[Na+]
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0611

RTECSNumber : YV7876000

TSCA : No

Documents