missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Sodium 2-Nitroaniline-4-sulfonate 98.0+%, TCI America™
$71.33 - $593.92
Chemical Identifiers
CAS | 5042-33-1 |
---|---|
Molecular Formula | C6H5N2NaO5S |
Molecular Weight (g/mol) | 240.165 |
MDL Number | MFCD00070599 |
InChI Key | SXTOGAIYFMTUAD-UHFFFAOYSA-M |
Synonym | 4-Amino-3-nitrobenzenesulfonic Acid Sodium Salt, 2-Nitroaniline-4-sulfonic Acid Sodium Salt, Sodium 4-Amino-3-nitrobenzenesulfonate |
PubChem CID | 165281 |
IUPAC Name | sodium;4-amino-3-nitrobenzenesulfonate |
SMILES | C1=CC(=C(C=C1S(=O)(=O)[O-])[N+](=O)[O-])N.[Na+] |
Chemical Identifiers
5042-33-1 | |
240.165 | |
SXTOGAIYFMTUAD-UHFFFAOYSA-M | |
165281 | |
C1=CC(=C(C=C1S(=O)(=O)[O-])[N+](=O)[O-])N.[Na+] |
C6H5N2NaO5S | |
MFCD00070599 | |
4-Amino-3-nitrobenzenesulfonic Acid Sodium Salt, 2-Nitroaniline-4-sulfonic Acid Sodium Salt, Sodium 4-Amino-3-nitrobenzenesulfonate | |
sodium;4-amino-3-nitrobenzenesulfonate |
Specifications
5042-33-1 | |
MFCD00070599 | |
4-Amino-3-nitrobenzenesulfonic Acid Sodium Salt, 2-Nitroaniline-4-sulfonic Acid Sodium Salt, Sodium 4-Amino-3-nitrobenzenesulfonate | |
C1=CC(=C(C=C1S(=O)(=O)[O-])[N+](=O)[O-])N.[Na+] | |
240.165 | |
240.16 | |
Crystalline Powder |
C6H5N2NaO5S | |
25 g | |
SXTOGAIYFMTUAD-UHFFFAOYSA-M | |
sodium;4-amino-3-nitrobenzenesulfonate | |
165281 | |
≥98.0% (HPLC) | |
Sodium 2-Nitroaniline-4-sulfonate |
Safety and Handling
EINECSNumber : (3)-2079
TSCA : Yes