missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Sodium 2-dibenzofuransulfonate hydrate, 98+%

$65.57 - $212.42

Chemical Identifiers

CAS 94600-19-8
Molecular Formula C12H7NaO4S·xH2O
Formula Weight 270.24 (Anhydrous)
MDL Number MFCD08061352
Beilstein 3795215
Synonym 2-Dibenzofuransulfonic acid sodium salt hydrate
Chemical Name or Material Sodium 2-dibenzofuransulfonate hydrate
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1334706 SDS Alfa Aesar™
L1334706
5g Each for $212.42
AAL1334703 SDS Alfa Aesar™
L1334703
1g Each for $65.57
Specifications Description & Specifications

Chemical Identifiers

94600-19-8
270.24 (Anhydrous)
3795215
Sodium 2-dibenzofuransulfonate hydrate
C12H7NaO4S·xH2O
MFCD08061352
2-Dibenzofuransulfonic acid sodium salt hydrate

Specifications

94600-19-8
270.24 (Anhydrous)
3795215
>300°C
Sodium 2-dibenzofuransulfonate hydrate
C12H7NaO4S·xH2O
MFCD08061352
2-Dibenzofuransulfonic acid sodium salt hydrate
5g
≥98%