missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Sodium 2-dibenzofuransulfonate hydrate, 98+%

Manufacturer:  Alfa Aesar™ L1334703

 View more versions of this product

Catalog No. AAL1334703



Specifications

Specifications

Sodium 2-dibenzofuransulfonate hydrate
1g
C12H9NaO5S
3795215
JKHBTMNKDUBHGC-UHFFFAOYSA-M
sodium;dibenzofuran-2-sulfonate;hydrate
57356836
≥98%
>300°C
94600-19-8
MFCD08061352
sodium 2-dibenzofuransulphonate hydrate, sodium 2-dibenzofuransulfonate hydrate, sodium dibenzo b,d furan-2-sulfonate-water 1/1/1, sodium 8-oxatricyclo 7.4.0.0 2 ,? trideca-1 13 ,2 7 ,3,5,9,11-hexaene-4-sulfonate hydrate, sodium 8-oxatricyclo 7.4.0.0 2 ,? trideca-1 9 ,2 7 ,3,5,10,12-hexaene-4-sulfonate hydrate, sodium 8-oxatricyclo 7.4.0.0_,? trideca-1 9 ,2 7 ,3,5,10,12-hexaene-4-sulfonate hydrate
C1=CC=C2C(=C1)C3=C(O2)C=CC(=C3)S(=O)(=O)[O-].O.[Na+]
288.249
270.24 (Anhydrous)

Chemical Identifiers

94600-19-8
288.249
JKHBTMNKDUBHGC-UHFFFAOYSA-M
57356836
C1=CC=C2C(=C1)C3=C(O2)C=CC(=C3)S(=O)(=O)[O-].O.[Na+]
C12H9NaO5S
MFCD08061352
sodium 2-dibenzofuransulphonate hydrate, sodium 2-dibenzofuransulfonate hydrate, sodium dibenzo b,d furan-2-sulfonate-water 1/1/1, sodium 8-oxatricyclo 7.4.0.0 2 ,? trideca-1 13 ,2 7 ,3,5,9,11-hexaene-4-sulfonate hydrate, sodium 8-oxatricyclo 7.4.0.0 2 ,? trideca-1 9 ,2 7 ,3,5,10,12-hexaene-4-sulfonate hydrate, sodium 8-oxatricyclo 7.4.0.0_,? trideca-1 9 ,2 7 ,3,5,10,12-hexaene-4-sulfonate hydrate
sodium;dibenzofuran-2-sulfonate;hydrate
Documents