Learn More
Sodium 2-bromoethanesulfonate, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1507622
Description
Sodium 2-bromoethanesulfonate(BES) was used to investigate the effect of BES on inhibition of bacterial growth.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsSodium 2-bromoethanesulfonate(BES) was used to investigate the effect of BES on inhibition of bacterial growth.
Solubility
Soluble in water and methanol.
Notes
Store in cool, dry place in tightly closed container. Store away from oxidizing agent.
Specifications
Sodium 2-bromoethanesulfonate | |
∼284°C (decomposition) | |
MFCD00007530 | |
4590828 | |
Soluble in water and methanol. | |
C(CBr)S(=O)(=O)[O-].[Na+] | |
211.005 | |
211.02 |
4263-52-9 | |
C2H4BrNaO3S | |
100 g | |
sodium 2-bromoethanesulfonate, 2-bromoethanesulfonic acid sodium salt, sodium 2-bromoethanesulphonate, 2-bromoethanesulfonic acid, sodium salt, sodium bromoethylsulfonate, ethanesulfonic acid, 2-bromo-, sodium salt, ethanesulfonic acid, 2-bromo-, sodium salt 1:1, sodium 2-bromoethane sulfonate, 2-bromoethanesulfonic acid sodium, natrium-2-bromethansulfonat | |
HNFOAHXBHLWKNF-UHFFFAOYSA-M | |
sodium;2-bromoethanesulfonate | |
23666797 | |
98% |
Chemical Identifiers
4263-52-9 | |
211.005 | |
HNFOAHXBHLWKNF-UHFFFAOYSA-M | |
23666797 | |
C(CBr)S(=O)(=O)[O-].[Na+] |
C2H4BrNaO3S | |
MFCD00007530 | |
sodium 2-bromoethanesulfonate, 2-bromoethanesulfonic acid sodium salt, sodium 2-bromoethanesulphonate, 2-bromoethanesulfonic acid, sodium salt, sodium bromoethylsulfonate, ethanesulfonic acid, 2-bromo-, sodium salt, ethanesulfonic acid, 2-bromo-, sodium salt 1:1, sodium 2-bromoethane sulfonate, 2-bromoethanesulfonic acid sodium, natrium-2-bromethansulfonat | |
sodium;2-bromoethanesulfonate |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 224-244-4
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only