missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Sodium 2-bromoethanesulfonate, 98%

Manufacturer:  Alfa Aesar™ A1507614

 View more versions of this product

Catalog No. AAA1507614



Description

Description

Applications
Sodium 2-bromoethanesulfonate(BES) was used to investigate the effect of BES on inhibition of bacterial growth.

Solubility
Soluble in water and methanol.

Notes
Store in cool, dry place in tightly closed container. Store away from oxidizing agent.
Specifications

Specifications

Sodium 2-bromoethanesulfonate
25g
C2H4BrNaO3S
4590828
HNFOAHXBHLWKNF-UHFFFAOYSA-M
sodium;2-bromoethanesulfonate
23666797
98%
~284°C (decomposition)
4263-52-9
MFCD00007530
sodium 2-bromoethanesulfonate, 2-bromoethanesulfonic acid sodium salt, sodium 2-bromoethanesulphonate, 2-bromoethanesulfonic acid, sodium salt, sodium bromoethylsulfonate, ethanesulfonic acid, 2-bromo-, sodium salt, ethanesulfonic acid, 2-bromo-, sodium salt 1:1, sodium 2-bromoethane sulfonate, 2-bromoethanesulfonic acid sodium, natrium-2-bromethansulfonat
C(CBr)S(=O)(=O)[O-].[Na+]
211.005
211.02

Chemical Identifiers

4263-52-9
211.005
HNFOAHXBHLWKNF-UHFFFAOYSA-M
23666797
C(CBr)S(=O)(=O)[O-].[Na+]
C2H4BrNaO3S
MFCD00007530
sodium 2-bromoethanesulfonate, 2-bromoethanesulfonic acid sodium salt, sodium 2-bromoethanesulphonate, 2-bromoethanesulfonic acid, sodium salt, sodium bromoethylsulfonate, ethanesulfonic acid, 2-bromo-, sodium salt, ethanesulfonic acid, 2-bromo-, sodium salt 1:1, sodium 2-bromoethane sulfonate, 2-bromoethanesulfonic acid sodium, natrium-2-bromethansulfonat
sodium;2-bromoethanesulfonate
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA

Documents