Please login to your online account to display your discounted pricing

Alfa Aesar™ Sodium 1-undecyl sulfate, 99% (dry wt.), water <2%

$84.53 - $302.86

Chemical Identifiers

Chemical Name or Material Sodium 1-undecyl sulfate
CAS 1072-24-8
Molecular Formula C11H23NaO4S
Linear Formula CH3(CH2)10OSO3Na
Formula Weight 274.35
MDL Number MFCD00053627
Synonym n-Undecyl sulfate sodium salt
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2214106

  SDS
alfa aesar™
B2214106
5g Each for $302.86

AAB2214103

  SDS
alfa aesar™
B2214103
1g Each for $84.53

AAB2214114

  SDS
alfa aesar™
B2214114
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Sodium 1-undecyl sulfate
CAS 1072-24-8
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Assay 99%
Assay Percent Notes (dry wt.), water <2%
Molecular Formula C11H23NaO4S
Linear Formula CH3(CH2)10OSO3Na
Formula Weight 274.35
MDL Number MFCD00053627
Synonym n-Undecyl sulfate sodium salt
Melting Point 208° to 210°C
Quantity 5g

Applications
Sodium 1-undecyl sulfate is used in the colorimetric measurements.

Solubility
Soluble in water.

Notes
Incompatible with strong oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.