missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium 1-Tridecanesulfonate 98.0+%, TCI America™

$266.63 - $752.17

Chemical Identifiers

CAS 5802-89-1
Molecular Formula C13H27NaO3S
Molecular Weight (g/mol) 286.406
MDL Number MFCD00024993
InChI Key CACJZDMMUHMEBN-UHFFFAOYSA-M
Synonym IPC-ALKS-13, 1-Tridecanesulfonic Acid Sodium Salt
PubChem CID 23683718
IUPAC Name sodium;tridecane-1-sulfonate
SMILES CCCCCCCCCCCCCS(=O)(=O)[O-].[Na+]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
I03515G View Documents Tci America
I03515G
5g Each for $266.63
 
I035125G View Documents Tci America
I035125G
25g Each for $752.17
 
Specifications

Chemical Identifiers

5802-89-1
286.406
CACJZDMMUHMEBN-UHFFFAOYSA-M
23683718
CCCCCCCCCCCCCS(=O)(=O)[O-].[Na+]
C13H27NaO3S
MFCD00024993
IPC-ALKS-13, 1-Tridecanesulfonic Acid Sodium Salt
sodium;tridecane-1-sulfonate

Specifications

White
5802-89-1
MFCD00024993
CACJZDMMUHMEBN-UHFFFAOYSA-M
sodium;tridecane-1-sulfonate
23683718
Crystal-Powder at 20°C
Sodium 1-Tridecanesulfonate [Reagent for Ion-Pair Chromatography]
5g
C13H27NaO3S
IPC-ALKS-13, 1-Tridecanesulfonic Acid Sodium Salt
CCCCCCCCCCCCCS(=O)(=O)[O-].[Na+]
286.406
286.41
≥98.0% (T)
Safety and Handling

Safety and Handling

EINECSNumber : (2)-1640

TSCA : No