missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Sodium 1-pentyl sulfate, 99%

$220.53 - $852.80

Chemical Identifiers

CAS 556-76-3
Molecular Formula C5H11NaO4S
Molecular Weight (g/mol) 190.189
MDL Number MFCD00041882
InChI Key KLZYTGLGWPHIDV-UHFFFAOYSA-M
Synonym n-amyl sulfate sodium salt
PubChem CID 23680281
IUPAC Name sodium;pentyl sulfate
SMILES CCCCCOS(=O)(=O)[O-].[Na+]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAB2242706 View Documents Alfa Aesar™
B2242706
5g Each for $220.53
 
AAB2242714 View Documents Alfa Aesar™
B2242714
25g Each for $852.80
 
Description

Description

Applications
Sodium 1-pentyl sulfate is used as a pharmaceutical intermediate, in chemical research.

Solubility
Soluble in water.

Notes
Store in cool, dry place in tightly closed containers. Store away from strong oxidizing agents.
Specifications

Chemical Identifiers

556-76-3
190.189
KLZYTGLGWPHIDV-UHFFFAOYSA-M
23680281
CCCCCOS(=O)(=O)[O-].[Na+]
C5H11NaO4S
MFCD00041882
n-amyl sulfate sodium salt
sodium;pentyl sulfate

Specifications

5g
C5H11NaO4S
4163608
KLZYTGLGWPHIDV-UHFFFAOYSA-M
sodium;pentyl sulfate
23680281
99%
556-76-3
MFCD00041882
n-amyl sulfate sodium salt
CCCCCOS(=O)(=O)[O-].[Na+]
190.189
190.2
Sodium 1-pentyl sulfate
Safety and Handling

Safety and Handling

TSCA : TSCA