Alfa Aesar™ Sodium 1-pentanesulfonate monohydrate, HPLC grade
Ion pair reagent
Manufacturer:  Alfa Aesar™ 1571406
 View more versions of this product
Catalog No. AA1571406
Specifications
Specifications
Sodium 1-pentanesulfonate monohydrate
5g
C5H13NaO4S
MFCD00007541
sodium pentane-1-sulfonate hydrate, 1-pentanesulfonic acid sodium salt monohydrate, sodium 1-pentanesulfonate monohydrate, potassium hydrate pentane-1-sulfonate, sodium 1-pentanesulfonate hydrate, sodium hydrate pentane-1-sulfonate, sodium pentane-1-sulfonate-water 1/1/1, 1-pentane sulphonic acid sodium salt monohydrate, 1-pentanesulfonic acid sodium salt monohydydrate, sodium 1-pentanesulfonate monohydrate, hplc grade
CCCCCS(=O)(=O)[O-].O.[Na+]
192.205
192.21 (174.19 Anhydrous)
HPLC
White
207605-40-1
CH3(CH2)4SO3Na·H2O
3725147
FPQYXAFKHLSWTI-UHFFFAOYSA-M
sodium;pentane-1-sulfonate;hydrate
23693099
Crystalline Powder
Chemical Identifiers
207605-40-1
192.205
FPQYXAFKHLSWTI-UHFFFAOYSA-M
23693099
CCCCCS(=O)(=O)[O-].O.[Na+]
C5H13NaO4S
MFCD00007541
sodium pentane-1-sulfonate hydrate, 1-pentanesulfonic acid sodium salt monohydrate, sodium 1-pentanesulfonate monohydrate, potassium hydrate pentane-1-sulfonate, sodium 1-pentanesulfonate hydrate, sodium hydrate pentane-1-sulfonate, sodium pentane-1-sulfonate-water 1/1/1, 1-pentane sulphonic acid sodium salt monohydrate, 1-pentanesulfonic acid sodium salt monohydydrate, sodium 1-pentanesulfonate monohydrate, hplc grade
sodium;pentane-1-sulfonate;hydrate
Safety and Handling
Safety and Handling
TSCA : TSCA
SDS Documents