Learn More
Sodium 1-pentanesulfonate monohydrate, HPLC grade, Thermo Scientific Chemicals
Ion pair reagent
Supplier: Thermo Scientific Chemicals 01571414
Description
Ion pair reagent.Sodium 1-pentanesulfonate monohydrate is used as a surfactant and a reversed-phase ion-pairing agent. It is utilized to study its effects on the properties of polyaniline, using Fourier transform infrared and UV-visible spectroscopy. Further, it is used for HPLC analysis of proteins and peptides.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Sodium 1-pentanesulfonate monohydrate | |
White | |
CH3(CH2)4SO3Na·H2O | |
25 g | |
sodium pentane-1-sulfonate hydrate, 1-pentanesulfonic acid sodium salt monohydrate, sodium 1-pentanesulfonate monohydrate, potassium hydrate pentane-1-sulfonate, sodium 1-pentanesulfonate hydrate, sodium hydrate pentane-1-sulfonate, sodium pentane-1-sulfonate-water 1/1/1, 1-pentane sulphonic acid sodium salt monohydrate, 1-pentanesulfonic acid sodium salt monohydydrate, sodium 1-pentanesulfonate monohydrate, hplc grade | |
FPQYXAFKHLSWTI-UHFFFAOYSA-M | |
sodium;pentane-1-sulfonate;hydrate | |
23693099 | |
HPLC |
207605-40-1 | |
C5H13NaO4S | |
MFCD00007541 | |
3725147 | |
Soluble in water. | |
CCCCCS(=O)(=O)[O-].O.[Na+] | |
192.205 | |
192.21 (174.19 Anhydrous) | |
Crystalline Powder |
Chemical Identifiers
207605-40-1 | |
192.205 | |
FPQYXAFKHLSWTI-UHFFFAOYSA-M | |
23693099 | |
CCCCCS(=O)(=O)[O-].O.[Na+] |
C5H13NaO4S | |
MFCD00007541 | |
sodium pentane-1-sulfonate hydrate, 1-pentanesulfonic acid sodium salt monohydrate, sodium 1-pentanesulfonate monohydrate, potassium hydrate pentane-1-sulfonate, sodium 1-pentanesulfonate hydrate, sodium hydrate pentane-1-sulfonate, sodium pentane-1-sulfonate-water 1/1/1, 1-pentane sulphonic acid sodium salt monohydrate, 1-pentanesulfonic acid sodium salt monohydydrate, sodium 1-pentanesulfonate monohydrate, hplc grade | |
sodium;pentane-1-sulfonate;hydrate |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 245-208-4
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only