Learn More
Sodium 1-octanesulfonate monohydrate, 99+%, Thermo Scientific Chemicals
$127.84 - $371.52
Chemical Identifiers
CAS | 207596-29-0 |
---|---|
Molecular Formula | C8H19NaO4S |
Molecular Weight (g/mol) | 234.29 |
MDL Number | MFCD00149551 |
InChI Key | MBURIAHQXJQKRE-UHFFFAOYSA-M |
Synonym | sodium octane-1-sulfonate hydrate, 1-octanesulfonic acid sodium salt monohydrate, sodium 1-octanesulfonate monohydrate, 1-octanesulfonic acid, sodium salt, monohydrate, sodium 1-octanesulfonate hydrate, 1-octane sulfonic acid, na, acmc-20p1dl, ksc496c2b, sodium1-octanesulfonatemonohydrate, potassium hydrate octane-1-sulfonate |
PubChem CID | 23666339 |
IUPAC Name | sodium;octane-1-sulfonate;hydrate |
SMILES | O.[Na+].CCCCCCCCS([O-])(=O)=O |
Description
Sodium 1-octanesulfonate monohydrate is used as an anionic detergent. It is an ion pairing reagent used in the HPLC analysis of proteins and peptides. Further, it is used as a surface active agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
207596-29-0 | |
234.29 | |
MBURIAHQXJQKRE-UHFFFAOYSA-M | |
23666339 | |
O.[Na+].CCCCCCCCS([O-])(=O)=O |
C8H19NaO4S | |
MFCD00149551 | |
sodium octane-1-sulfonate hydrate, 1-octanesulfonic acid sodium salt monohydrate, sodium 1-octanesulfonate monohydrate, 1-octanesulfonic acid, sodium salt, monohydrate, sodium 1-octanesulfonate hydrate, 1-octane sulfonic acid, na, acmc-20p1dl, ksc496c2b, sodium1-octanesulfonatemonohydrate, potassium hydrate octane-1-sulfonate | |
sodium;octane-1-sulfonate;hydrate |
Specifications
207596-29-0 | |
C8H19NaO4S | |
10 g | |
Soluble in water. | |
O.[Na+].CCCCCCCCS([O-])(=O)=O | |
234.29 | |
234.29 (216.28 Anhydrous) | |
Crystalline |
>300°C | |
MFCD00149551 | |
sodium octane-1-sulfonate hydrate, 1-octanesulfonic acid sodium salt monohydrate, sodium 1-octanesulfonate monohydrate, 1-octanesulfonic acid, sodium salt, monohydrate, sodium 1-octanesulfonate hydrate, 1-octane sulfonic acid, na, acmc-20p1dl, ksc496c2b, sodium1-octanesulfonatemonohydrate, potassium hydrate octane-1-sulfonate | |
MBURIAHQXJQKRE-UHFFFAOYSA-M | |
sodium;octane-1-sulfonate;hydrate | |
23666339 | |
≥99% | |
Sodium 1-octanesulfonate monohydrate |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 226-195-4
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only