missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Sodium 1-Naphthaleneacetate, TCI America™

$30.56 - $252.09

Chemical Identifiers

CAS 61-31-4
Molecular Formula C12H9NaO2
Molecular Weight (g/mol) 208.192
MDL Number MFCD00064967
InChI Key CJUUXVFWKYRHAR-UHFFFAOYSA-M
Synonym 1-Naphthylacetic Acid Sodium Salt, 1-Naphthaleneacetic Acid Sodium Salt, Sodium 1-Naphthylacetate, Na-NAA
PubChem CID 23694671
ChEBI CHEBI:81811
IUPAC Name sodium;2-naphthalen-1-ylacetate
SMILES C1=CC=C2C(=C1)C=CC=C2CC(=O)[O-].[Na+]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
N000725G View Documents TCI America
N000725G
25g Each for $30.56
 
N0007500G View Documents TCI America
N0007500G
500g Each for $252.09
 
Specifications

Chemical Identifiers

61-31-4
208.192
CJUUXVFWKYRHAR-UHFFFAOYSA-M
23694671
sodium;2-naphthalen-1-ylacetate
C12H9NaO2
MFCD00064967
1-Naphthylacetic Acid Sodium Salt, 1-Naphthaleneacetic Acid Sodium Salt, Sodium 1-Naphthylacetate, Na-NAA
CHEBI:81811
C1=CC=C2C(=C1)C=CC=C2CC(=O)[O-].[Na+]

Specifications

White-Yellow
61-31-4
MFCD00064967
CJUUXVFWKYRHAR-UHFFFAOYSA-M
sodium;2-naphthalen-1-ylacetate
23694671
208.19
≥96.0% (HPLC,T)
25g
C12H9NaO2
1-Naphthylacetic Acid Sodium Salt, 1-Naphthaleneacetic Acid Sodium Salt, Sodium 1-Naphthylacetate, Na-NAA
C1=CC=C2C(=C1)C=CC=C2CC(=O)[O-].[Na+]
208.192
CHEBI:81811
Crystal-Powder at 20°C
Sodium 1-Naphthaleneacetate
Safety and Handling

Safety and Handling

EINECSNumber : (4)-0410

RTECSNumber : QJ1355000

TSCA : Yes