missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Sodium 1-hexanesulfonate monohydrate, 99%

Manufacturer:  Alfa Aesar™ 3219009

 View more versions of this product

Catalog No. AA3219009



Specifications

Specifications

Sodium 1-hexanesulfonate monohydrate
10g
C6H15NaO4S
MFCD00149549
1-hexanesulfonic acid sodium salt monohydrate, sodium hexane-1-sulfonate hydrate, sodium 1-hexanesulfonate monohydrate, n-1-hexanesulfonic acid, sodium 1-hexanesulfonate hydrate, sodium n-hexanesulfonate monohydrate, potassium hexane-1-sulfonate hydrate, sodium hexane-1-sulphonate monohydrate, sodium hexane-1-sulfonate-water 1/1/1, 1-hexanesulfonic acid,sodium salt, hydrate 1:1:1
CCCCCCS(=O)(=O)[O-].O.[Na+]
206.232
206.24 (188.22 Anhydrous)
99%
White
207300-91-2
CH3(CH2)5SO3Na·H2O
3727014
CLCGFJYKZGFGSQ-UHFFFAOYSA-M
sodium;hexane-1-sulfonate;hydrate
23696962
Crystalline

Chemical Identifiers

207300-91-2
206.232
CLCGFJYKZGFGSQ-UHFFFAOYSA-M
23696962
CCCCCCS(=O)(=O)[O-].O.[Na+]
C6H15NaO4S
MFCD00149549
1-hexanesulfonic acid sodium salt monohydrate, sodium hexane-1-sulfonate hydrate, sodium 1-hexanesulfonate monohydrate, n-1-hexanesulfonic acid, sodium 1-hexanesulfonate hydrate, sodium n-hexanesulfonate monohydrate, potassium hexane-1-sulfonate hydrate, sodium hexane-1-sulphonate monohydrate, sodium hexane-1-sulfonate-water 1/1/1, 1-hexanesulfonic acid,sodium salt, hydrate 1:1:1
sodium;hexane-1-sulfonate;hydrate
Safety and Handling

Safety and Handling

TSCA : TSCA

SDS Documents