Learn More
Sodium 1-heptanesulfonate monohydrate, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 01521409
Description
Sodium 1-heptanesulfonate monohydrate is an ion pairing reagent used in the HPLC analysis of proteins and peptides. It is utilized in high performance capillary electrophoresis analysis of peptides.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Sodium 1-heptanesulfonate monohydrate | |
>300°C | |
C7H17NaO4S | |
MFCD00149550 | |
3731762 | |
Slightly soluble in water. | |
CCCCCCCS(=O)(=O)[O-].O.[Na+] | |
220.259 | |
220.26 (202.24 Anhydrous) | |
Crystalline |
207300-90-1 | |
White | |
CH3(CH2)6SO3Na·H2O | |
10 g | |
sodium heptane-1-sulfonate hydrate, 1-heptanesulfonic acid sodium salt monohydrate, sodium 1-heptanesulfonate monohydrate, sodium 1-heptanesulfonate hydrate, sodium1-heptanesulfonatemonohydrate, potassium heptane-1-sulfonate hydrate, sodium heptane-1-sulfonate-water 1/1/1, sodium 1-heptanesulfonate monohydrate t, sodium 1-heptanesulfonate monohydrate, for hplc | |
XWZCREJRXRKIRQ-UHFFFAOYSA-M | |
sodium;heptane-1-sulfonate;hydrate | |
23687711 | |
99% |
Chemical Identifiers
207300-90-1 | |
220.259 | |
XWZCREJRXRKIRQ-UHFFFAOYSA-M | |
23687711 | |
CCCCCCCS(=O)(=O)[O-].O.[Na+] |
C7H17NaO4S | |
MFCD00149550 | |
sodium heptane-1-sulfonate hydrate, 1-heptanesulfonic acid sodium salt monohydrate, sodium 1-heptanesulfonate monohydrate, sodium 1-heptanesulfonate hydrate, sodium1-heptanesulfonatemonohydrate, potassium heptane-1-sulfonate hydrate, sodium heptane-1-sulfonate-water 1/1/1, sodium 1-heptanesulfonate monohydrate t, sodium 1-heptanesulfonate monohydrate, for hplc | |
sodium;heptane-1-sulfonate;hydrate |
Safety and Handling
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 245-210-5
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only