missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium 1-Decanesulfonate 98.0+%, TCI America™
Supplier: TCI America D11815G
Specifications
| Sodium 1-Decanesulfonate | |
| White | |
| MFCD00007526 | |
| sodium decane-1-sulfonate, sodium 1-decanesulfonate, 1-decanesulfonic acid sodium salt, 1-decanesulfonic acid, sodium salt, decyl sodium sulfonate, sodium decane-1-sulphonate, ipc-alks-10, n-decylsulfonic acid, sodium salt, 1-decanesulphonic acid, sodium salt, 2-decanesulfonic acid, sodium salt | |
| CCCCCCCCCCS(=O)(=O)[O-].[Na+] | |
| 244.325 | |
| 244.32 | |
| Crystalline Powder |
| 13419-61-9 | |
| C10H21NaO3S | |
| 5 g | |
| AIMUHNZKNFEZSN-UHFFFAOYSA-M | |
| sodium;decane-1-sulfonate | |
| 2724181 | |
| ≥98.0% (T) |
Chemical Identifiers
| 13419-61-9 | |
| 244.325 | |
| AIMUHNZKNFEZSN-UHFFFAOYSA-M | |
| 2724181 | |
| CCCCCCCCCCS(=O)(=O)[O-].[Na+] |
| C10H21NaO3S | |
| MFCD00007526 | |
| sodium decane-1-sulfonate, sodium 1-decanesulfonate, 1-decanesulfonic acid sodium salt, 1-decanesulfonic acid, sodium salt, decyl sodium sulfonate, sodium decane-1-sulphonate, ipc-alks-10, n-decylsulfonic acid, sodium salt, 1-decanesulphonic acid, sodium salt, 2-decanesulfonic acid, sodium salt | |
| sodium;decane-1-sulfonate |
Safety and Handling
EINECSNumber : (2)-1640
TSCA : No