missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Soda lime, indicating, ACS, Alfa Aesar™

Manufacturer:  Alfa Aesar 4469736

 View more versions of this product

Catalog No. AA4469736



Specifications

Specifications

Soda lime
8006-28-8
CaHNaO2
MFCD00134094
Odorless
Air Sensitive & Hygroscopic
[OH-].O=[Ca].[Na+]
96.074
4 to 8 mesh
Indicating
500g
4 to 8 Mesh
UN1907
14,8569
HUAUNKAZQWMVFY-UHFFFAOYSA-M
sodium;oxocalcium;hydroxide
66545795
ACS

Chemical Identifiers

8006-28-8
96.074
HUAUNKAZQWMVFY-UHFFFAOYSA-M
sodium;oxocalcium;hydroxide
CaHNaO2
MFCD00134094
66545795
[OH-].O=[Ca].[Na+]
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: SODA LIME

TSCA : TSCA

SDS Documents