Learn More
Soda lime, ACS, Thermo Scientific Chemicals
$111.25 - $424.49
Chemical Identifiers
CAS | 8006-28-8 |
---|---|
Molecular Formula | CaHNaO2 |
Molecular Weight (g/mol) | 96.074 |
MDL Number | MFCD00134094 |
InChI Key | HUAUNKAZQWMVFY-UHFFFAOYSA-M |
PubChem CID | 66545795 |
IUPAC Name | sodium;oxocalcium;hydroxide |
SMILES | [OH-].O=[Ca].[Na+] |
Description
Soda lime's ability to absorb carbon dioxide makes it valuable in medical and surgical professions. It is also used as powerful drying agent, or desiccant, in commercial and industrial usage. It is also useful for the construction of gas masks. It plays an essential role for the measurement of soil CO2 efflux upon absorption. It is also useful for direct measurement of dissolved CO2.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
8006-28-8 | |
96.074 | |
HUAUNKAZQWMVFY-UHFFFAOYSA-M | |
sodium;oxocalcium;hydroxide |
CaHNaO2 | |
MFCD00134094 | |
66545795 | |
[OH-].O=[Ca].[Na+] |
Specifications
4-8 Mesh | |
Soda Lime | |
CaHNaO2 | |
4 to 8 | |
HUAUNKAZQWMVFY-UHFFFAOYSA-M | |
Air sensitive; Hygroscopic | |
96.074 | |
ACS Reagent |
8006-28-8 | |
500 g | |
MFCD00134094 | |
Soluble in water. | |
[OH-].O=[Ca].[Na+] | |
sodium;oxocalcium;hydroxide | |
66545795 | |
Soda Lime |
Safety and Handling
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P390-P501c
H314-H335
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only