Please login to your online account to display your discounted pricing

Sod Pot Tartrate,0.25% (w/v), Ricca Chemical


Quantity 20L

Manufacturer: ricca chemical company  RIR753500020F

 View more versions of this product

Catalog No. R753500020F

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Quantity 20L
Packaging Cubitainer
Assay Percent Range 0.25% (w/v)
Chemical Name or Material Sod Pot Tartrate

Specifications

Chemical Name or Material Sod Pot Tartrate
Assay Percent Range 0.25% (w/v)