Please login to your online account to display your discounted pricing

Sod Pot Tartrate,0.25% (w/v), Ricca Chemical


Quantity 1L

Manufacturer: ricca chemical company  RIR75350001A

 View more versions of this product

Catalog No. R75350001A

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Quantity 1L
Chemical Name or Material Sod Pot Tartrate
Packaging Poly Bottle
Assay Percent Range 0.25% (w/v)

Specifications

Chemical Name or Material Sod Pot Tartrate
Assay Percent Range 0.25% (w/v)